Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDKL1 Monoclonal Antibody (OTI5B10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA805952

Description
This gene product is a member of a large family of CDC2-related serine/threonine protein kinases. It accumulates primarily in the nucleus. Alternative splice variants encoding different protein isoforms have been described, but their full-length nature has not been determined.Specifications
CDKL1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q00532 | |
CDKL1 | |
Human recombit protein fragment corresponding to amino acids 152-357 of human CDKL1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI5B10 | |
Unconjugated | |
CDKL1 | |
KKIALRE, P42 | |
Mouse | |
Affinity Chromatography | |
RUO | |
8814 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction