Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDKL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545998 Supplier Invitrogen™ Supplier No. PA545998
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EKYEKIGKIG EGSYGVVFKC RNRDTGQIVA IKKFLESEDD PVIKKIALRE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

This gene product is a member of a large family of CDC2-related serine/threonine protein kinases. It accumulates primarily in the nucleus. Alternative splice variants encoding different protein isoforms have been described, but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDKL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CDKL1
Gene Accession No. Q00532
Gene Alias 4933411O17Rik; CDC2-related kinase 1; Cdkl1; cyclin dependent kinase like 1; cyclin-dependent kinase-like 1; cyclin-dependent kinase-like 1 (CDC2-related kinase); KKIALRE; P42; protein kinase p42 KKIALRE; serine/threonine protein kinase KKIALRE; serine/threonine-protein kinase KKIALRE; zgc:101002
Gene Symbols CDKL1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CDKL1 (aa 3-52).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8814
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.