Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDKL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566809
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566809 Supplier Invitrogen™ Supplier No. PA566809
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLRKSRKHHCF TETSKLQYLP QLTGSSILPA LDNKKYYCDT KKLNY Highest antigen sequence identity to the following orthologs - mouse 67%, rat 61%.

This gene product is a member of a large family of CDC2-related serine/threonine protein kinases. It accumulates primarily in the nucleus. Alternative splice variants encoding different protein isoforms have been described, but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDKL1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDKL1
Gene Accession No. Q00532
Gene Alias 4933411O17Rik; CDC2-related kinase 1; Cdkl1; cyclin dependent kinase like 1; cyclin-dependent kinase-like 1; cyclin-dependent kinase-like 1 (CDC2-related kinase); KKIALRE; P42; protein kinase p42 KKIALRE; serine/threonine protein kinase KKIALRE; serine/threonine-protein kinase KKIALRE; zgc:101002
Gene Symbols CDKL1
Host Species Rabbit
Immunogen Recombinant Human CDKL1. Recombinant protein control fragment (Product #RP-107465).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8814
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.