Learn More
Invitrogen™ CDKL2 Monoclonal Antibody (OTI5D9)

Description
CDKL2 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | CDKL2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI5D9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CDKL2 |
| Gene Accession No. | Q92772 |
| Gene Alias | 5330436L21Rik; AI505225; CDC2-related kinase; Cdkl2; cyclin dependent kinase like 2; cyclin-dependent kinase-like 2; cyclin-dependent kinase-like 2 (CDC2-related kinase); Kkiamre; Kkm; P56; p56 KKIAMRE protein kinase; protein kinase p56 KKIAMRE; serine/threonine protein kinase KKIAMRE; serine/threonine-protein kinase KKIAMRE; testicular tissue protein Li 35 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.