Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDKL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104539
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104539 Supplier Invitrogen™ Supplier No. PA5104539
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CDKL4.

CDKL4 (Cyclin Dependent Kinase Like 4) is a Protein Coding gene. Diseases associated with CDKL4 include Orofaciodigital Syndrome Vi. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is CDKL1. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDKL4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDKL4
Gene Accession No. Q5MAI5
Gene Alias AU067824; CDKL4; cyclin dependent kinase like 4; cyclin-dependent kinase-like 4; EC 2.7.11.22; Gm942
Gene Symbols CDKL4
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDKL4(Accession Q5MAI5), corresponding to amino acid residues D273-T323.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 344387, 503009
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.