Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKL5 Antibody, ProSci
SDP

Catalog No. 16591085
Change view
Click to view available options
Quantity:
0.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-591-085 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-591-085 Supplier ProSci LLC Supplier No. 627570400
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDKL5 Polyclonal Antibody specifically detects CASC3 in Human, Mouse Samples. It is validated for Western Blot, Immunohistochemistry (Paraffin).

Specifications

Antigen CDKL5
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution For WB starting dilution is: 1:1000, For IHC-P starting dilution is: 1:10~50
Formulation Supplied in PBS with 0.09% (W/V) sodium azide.
Gene Accession No. O76039
Gene Alias Cyclin-dependent kinase-like 5, Serine/threonine-protein kinase 9, CDKL5, STK9
Host Species Rabbit
Immunogen This CDKL5 (STK9) antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 982-1012 amino acids from the C-terminal region of human CDKL5 (STK9).
Purification Method This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
Quantity 0.4 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6792
Target Species Human, Mouse
Content And Storage Store at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Antibody
Form Liquid
Isotype Ig
Research Category Neuroscience, Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.