Learn More
Invitrogen™ CDKL5 Monoclonal Antibody (CL4888)

Description
Immunogen sequence: AARANSLQLL SPQPGEQLPP EMTVARSSVK ETSREGTSSF HTRQKSEGGV YHDPHSDDGT APKENRHLYN DPVPRRVGSF YRVPSPRPDN SFHENNVSTR VSSLPSESSS GTNHSKRQPA FDP Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 97%.
Specifications
Specifications
| Antigen | CDKL5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | CL4888 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CDKL5 |
| Gene Accession No. | O76039 |
| Gene Alias | BC038161; Cdkl5; CFAP247; cyclin dependent kinase 5 transcript; cyclin dependent kinase like 5; cyclin-dependent kinase-like 5; EIEE2; ISSX; RP1-245G19.3; serine/threonine kinase 9; serine/threonine-protein kinase 9; STK9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.