Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDKL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA514083
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA514083 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA514083 Supplier Invitrogen™ Supplier No. PA514083
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDKL5 Polyclonal Antibody for Western Blot, IHC (P)

CDKL5 serine/threonine kinase (STK9) is associated with X-linked infantile spasm syndrome (ISSX), also known as X-linked West syndrome and Rett syndrome (RTT). CDKL5 can control nuclear speckle morphology by regulating the phosphorylation state of splicing regulatory proteins, and may be involved indirectly in pre-mRNA processing by controlling splicing factor dynamics. The mutations in CDKL5 are major cause of infantile spasms and early epileptic seizures in female patients, and of a later intractable seizure disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDKL5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CDKL5
Gene Accession No. O76039
Gene Alias BC038161; Cdkl5; CFAP247; cyclin dependent kinase 5 transcript; cyclin dependent kinase like 5; cyclin-dependent kinase-like 5; EIEE2; ISSX; RP1-245G19.3; serine/threonine kinase 9; serine/threonine-protein kinase 9; STK9
Gene Symbols CDKL5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 982-1012 amino acids from the C-terminal region of human CDKL5 (STK9).
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6792
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.