Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKL5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981456
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1456 Supplier Bioss Supplier No. BS11538R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Defects in STK9, a dual-specificity serine/threonine kinase, are a cause of atypical Rett syndrome. Rett syndrome is an X-linked dominant disease. It is a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females. Patients appear to develop normally until 6 to 18 months of age, then gradually lose speech and purposeful hand movements and develop microcephaly, seizures, autism, ataxia, intermittent hyperventilation, and stereotypic hand movements. After initial regression, the condition stabilizes and patients usually survive into adulthood. Rett syndrome due to CDKL5-associated mutations is characterized by a severe early-onset phenotype and atypical features such as infantile spasms.

Specifications

Antigen CDKL5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CDKL5
Gene Accession No. O76039
Gene Alias BC038161; CDKL5; CFAP247; cyclin dependent kinase 5 transcript; cyclin dependent kinase like 5; Cyclin-dependent kinase-like 5; EIEE2; ISSX; RP1-245G19.3; serine/threonine kinase 9; Serine/threonine-protein kinase 9; STK9
Gene Symbols CDKL5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CDKL5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6792
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.