Learn More
CDKN1A Mouse anti-Human, Clone: CIP1/823, Abnova™
Mouse monoclonal antibody raised against full length recombinant human CDKN1A.
Supplier: Abnova Corporation MAB13242
Description
This gene encodes a potent cyclin-dependent kinase inhibitor. The encoded protein binds to and inhibits the activity of cyclin-CDK2 or -CDK4 complexes, and thus functions as a regulator of cell cycle progression at G1. The expression of this gene is tightly controlled by the tumor suppressor protein p53, through which this protein mediates the p53-dependent cell cycle G1 phase arrest in response to a variety of stress stimuli. This protein can interact with proliferating cell nuclear antigen (PCNA), a DNA polymerase accessory factor, and plays a regulatory role in S phase DNA replication and DNA damage repair. This protein was reported to be specifically cleaved by CASP3-like caspases, which thus leads to a dramatic activation of CDK2, and may be instrumental in the execution of apoptosis following caspase activation. Two alternatively spliced variants, which encode an identical protein, have been reported. [provided by RefSeq]
Specifications
CDKN1A | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined | |
CAP20/CDKN1/CIP1/MDA-6/P21/SDI1/WAF1/p21CIP1 | |
Mouse | |
Protein A/G purification | |
RUO | |
1026 | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
CIP1/823 | |
Mouse monoclonal antibody raised against full length recombinant human CDKN1A. | |
In 10mM PBS. | |
CDKN1A | |
Recombinant protein corresponding to full length human CDKN1A. | |
100 μg | |
Primary | |
This monoclonal antibody is highly specific to CDKN1A and shows no cross-reaction with other closely related mitotic inhibitors. | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.