Learn More
CDKN1A Rabbit anti-Human, Unconjugated, Clone: CIP1/2489R, Abnova™
Shop All Abnova Corporation ProductsDescription
Specifications
Specifications
| Antigen | CDKN1A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CIP1/2489R |
| Conjugate | Unconjugated |
| Description | cyclin-dependent kinase inhibitor 1A (p21, Cip1) |
| Gene | CDKN1A |
| Gene Alias | CAP20, CDKN1, CIP1, MDA-6, P21, SDI1, WAF1, p21CIP1 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein corresponding to full-length human CDKN1A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.