Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdkn1b Mouse anti-Human, Mouse, Clone: DCS-72.F6, Abnova™

Catalog No. 89034304 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-304 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-304 Supplier Abnova Corporation Supplier No. MAB14203
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant mouse Cdkn1b.

  • High specificity
  • High reactivity

Specifications

Antigen Cdkn1b
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone DCS-72.F6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant mouse Cdkn1b.
Dilution Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determin
Formulation In PBS.
Gene Accession No. P46414
Gene Alias AA408329/AI843786/Kip1/p27/p27Kip1
Gene Symbols Cdkn1b
Host Species Mouse
Immunogen Recombinant protein corresponding to full length mouse Cdkn1b.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12576
Test Specificity The epitope of this antibody spans between amino acids 83-204 of mouse Cdkn1b. This antibody is highly specific and shows no cross-reaction with other related mitotic inhibitors.
Target Species Human, Mouse
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.