Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKN1C Mouse anti-Human, Clone: KP10, Abnova™

Catalog No. 89933938 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-938 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-938 Supplier Abnova Corporation Supplier No. MAB13248
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant human CDKN1C.

The protein encoded by this gene is a tight-binding, strong inhibitor of several G1 cyclin/Cdk complexes and a negative regulator of cell proliferation. Mutations in this gene are implicated in sporadic cancers and Beckwith-Wiedemann syndorome, suggesting that this gene is a tumor suppressor candidate. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen CDKN1C
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone KP10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human CDKN1C.
Dilution Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS.
Gene Alias BWCR/BWS/KIP2/WBS/p57
Gene Symbols CDKN1C
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human CDKN1C.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1028
Test Specificity This monoclonal antibody shows no cross-reaction with CDKN1B.
Target Species Human
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.