Learn More
Invitrogen™ CDKN2A (Isoform 3) Polyclonal Antibody

Description
Additional Information: This antibody is expected to recognize the pancreas isoform (NP_478104.1) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
Specifications
Specifications
| Antigen | CDKN2A (Isoform 3) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | CDKN2A |
| Gene Accession No. | P42771, Q8N726 |
| Gene Symbols | CDKN2A |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.