Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDKN2AIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541870
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541870 Supplier Invitrogen™ Supplier No. PA541870
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AIEALKATLD VFFVPLKELA DLPQNKSSQE SIVCELRCKS VYLGTGCGKS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

CARF was first cloned as a novel binding partner of ARF from a yeast-interactive screen. CARF and ARF colocalize in the perinucleolar region and have a collaborative function. In the nucleoplasm, CARF interacts with p53 and enhances its function. The p53 downregulates CARF in a negative feedback regulatory loop and may also involve p53 antagonist HDM2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDKN2AIP
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Cdkn2aip
Gene Accession No. Q9NXV6
Gene Alias 4921511I16Rik; AW208986; CARF; CDKN2A interacting protein; CDKN2A-interacting protein; CDKN2A-interacting protein-like protein; Cdkn2aip; CDKN2AIP-like protein; collaborates/cooperates with ARF (alternate reading frame) protein; collaborator of ARF; MGC137334 protein; RGD1305302
Gene Symbols Cdkn2aip
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CARF (aa 470-519).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55602
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.