Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDKN2AIP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566460
Description
Immunogen sequence: WANHVFLGCRY PQKVMDKILS MAEGIKVTDA PTYTTRDELV AKVKKRGISS SNEGVEEPSK KRVIEGKNSS AVEQDHAK Highest antigen sequence identity to the following orthologs - mouse 90%, rat 91%.
CARF was first cloned as a novel binding partner of ARF from a yeast-interactive screen. CARF and ARF colocalize in the perinucleolar region and have a collaborative function. In the nucleoplasm, CARF interacts with p53 and enhances its function. The p53 downregulates CARF in a negative feedback regulatory loop and may also involve p53 antagonist HDM2.
Specifications
CDKN2AIP | |
Polyclonal | |
Unconjugated | |
Cdkn2aip | |
4921511I16Rik; AW208986; CARF; CDKN2A interacting protein; CDKN2A-interacting protein; CDKN2A-interacting protein-like protein; Cdkn2aip; CDKN2AIP-like protein; collaborates/cooperates with ARF (alternate reading frame) protein; collaborator of ARF; MGC137334 protein; RGD1305302 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55602 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NXV6 | |
Cdkn2aip | |
Recombinant Human CDKN2AIP. Recombinant protein control fragment (Product #RP-107135). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction