Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDKN2AIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109507
Description
Immunogen sequence: MAQEVSEYLS QNPRVAAWVE ALRCDGETDK HWRHRRDFLL RNAGDLAPAG GAASASTDEA ADAESGTRNR QLQQLISFSM AWANHVFLGC RYPQKVMDKI LSMAEGIKVT DAPTYTTRDE LVAKVKKRGI SSSNEGVEEP SKKRVIEGKN SSAVEQDHAK TSAKTERASA QQENSSTCIG SAIKSESGNS ARSSGISSQN SSTSDGDRSV SSQSSSSVSS.
CARF was first cloned as a novel binding partner of ARF from a yeast-interactive screen. CARF and ARF colocalize in the perinucleolar region and have a collaborative function. In the nucleoplasm, CARF interacts with p53 and enhances its function. The p53 downregulates CARF in a negative feedback regulatory loop and may also involve p53 antagonist HDM2.
Specifications
| CDKN2AIP | |
| Polyclonal | |
| Unconjugated | |
| Cdkn2aip | |
| 4921511I16Rik; AW208986; CARF; CDKN2A interacting protein; CDKN2A-interacting protein; CDKN2A-interacting protein-like protein; Cdkn2aip; CDKN2AIP-like protein; collaborates/cooperates with ARF (alternate reading frame) protein; collaborator of ARF; MGC137334 protein; RGD1305302 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 306455, 55602, 70925 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.58 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5U2X0, Q8BI72, Q9NXV6 | |
| Cdkn2aip | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human CDKN2AIP (NP_060102.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction