Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKN2AIP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986023
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6023 Supplier Bioss Supplier No. BS4267R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CARF was first cloned as a novel binding partner of ARF from a yeast-interactive screen. CARF and ARF colocalize in the perinucleolar region and have a collaborative function. In the nucleoplasm, CARF interacts with p53 and enhances its function. The p53 downregulates CARF in a negative feedback regulatory loop and may also involve p53 antagonist HDM2.

Specifications

Antigen CDKN2AIP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Cdkn2aip
Gene Accession No. Q9NXV6
Gene Alias 4921511I16Rik; AW208986; BOS_23773; Carf; CDKN2A interacting protein; CDKN2A-interacting protein; CDKN2AIP; collaborates/cooperates with ARF (alternate reading frame) protein; Collaborator of ARF; LOC100340987; RGD1305302
Gene Symbols Cdkn2aip
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CARF.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55602
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.