Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKN2B Monoclonal Antibody (OTI7C4), TrueMAB™, OriGene
SDP

Catalog No. 502539850
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9850 30 μL
50-167-5731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9850 Supplier OriGene Supplier No. TA505205S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDKN2B Monoclonal antibody specifically detects CDKN2B in Human samples. It is validated for Western Blot

Specifications

Antigen CDKN2B
Applications Western Blot
Classification Monoclonal
Clone OTI7C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene OTUB1
Gene Accession No. Q96FW1
Gene Alias AV083695; CDK inhibitory protein; CDK4B inhibitor; CDK4I; Cdkn2b; cyclin dependent kinase inhibitor 2B; Cyclin dependent kinase inhibitor 2B (p15, inhibits CDK4); cyclin-dependent kinase 4 inhibitor B; cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4); cyclin-dependent kinase inhibitor p15; cyclin-dependent kinase inhibitor p15INK4b; cyclin-dependent kinase inhibitor protein; cyclin-dependent kinases 4 and 6 binding protein; Ink4; INK4B; MTS2; MTS-2; Multiple tumor suppressor 2; p14_CDK inhibitor; p14_INK4B; p14-INK4b; P15; p15 CDK inhibitor; p15(INK4b); p15_INK4B; p15INK4b; p15-INK4b; TP15
Host Species Mouse
Immunogen Full length human recombinant protein of human OTUB1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55611
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.