Learn More
CDKN2B Mouse anti-Human, Clone: OTI8H4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505208

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The four known 15-to 19 kDa INK4 proteins (p15INK4b, p16INK4a, p18INK4c andp19INK4d) bind and inhibit CDK4 and CDK6 but not other CDKs, including cyclin E-Cdk2, cyclin A-Cdk2 and cyclin B-Cdk1. Like the three D-type cyclins, the INK4 genes are expressed in distinct tissue-specific patterns, suggesting that they are not strictly redundant. 5 p18INK4c is expressed during G1 to S transition in the eukaryotic cell division cycle. It is maximally induced as cells enter S-phase.Specifications
CDKN2B | |
Monoclonal | |
Unconjugated | |
CDKN2B | |
CDK4I, INK4B, MTS2, P15, TP15, p15INK4b | |
Mouse | |
Affinity Chromatography | |
RUO | |
1030 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI8H4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P42772 | |
CDKN2B | |
Full length human recombit protein of human CDKN2B produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.