Learn More
CDKN2C Mouse anti-Human, Clone: OTI6B3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF801187

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the INK4 family of cyclin-dependent kinase inhibitors. This protein has been shown to interact with CDK4 or CDK6, and prevent the activation of the CDK kinases, thus function as a cell growth regulator that controls cell cycle G1 progression. Ectopic expression of this gene was shown to suppress the growth of human cells in a manner that appears to correlate with the presence of a wild-type RB1 function. Studies in the knockout mice suggested the roles of this gene in regulating spermatogenesis, as well as in suppressing tumorigenesis. Two alternatively spliced transcript variants of this gene, which encode an identical protein, have been reported.Specifications
CDKN2C | |
Monoclonal | |
Unconjugated | |
CDKN2C | |
INK4C, p18, p18-INK4C | |
Mouse | |
Affinity Chromatography | |
RUO | |
1031 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6B3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P42773 | |
CDKN2C | |
Full length human recombit protein of human CDKN2C produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.