Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDKN2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578378
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578378 Supplier Invitrogen™ Supplier No. PA578378
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Neuro2A, Raw264.7, C2C12, NIH-3T3 Predicted Reactivity: Human (100%), Rat (88%), Rhesus Monkey (100%), Bovine (89%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the INK4 family of cyclin-dependent kinase inhibitors. This protein has been shown to interact with CDK4 or CDK6, and prevent the activation of the CDK kinases, thus function as a cell growth regulator that controls cell cycle G1 progression. Ectopic expression of this gene was shown to suppress the growth of human cells in a manner that appears to correlate with the presence of a wild-type RB1 function. Studies in the knockout mice suggested the roles of this gene in regulating spermatogenesis, as well as in suppressing tumorigenesis. Two alternatively spliced transcript variants of this gene, which encode an identical protein, have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDKN2C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene CDKN2C
Gene Accession No. P42773, Q60772
Gene Alias C77269; CDK6 inhibitor p18; Cdkn2c; CDKN6; cyclin dependent kinase inhibitor 2C; cyclin-dependent inhibitor; cyclin-dependent kinase 4 inhibitor C; Cyclin-dependent kinase 6 inhibitor; cyclin-dependent kinase 6 inhibitor p18; cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4); INK4c; p18; p18INK4c; p18-INK4c; p18-INK6
Gene Symbols CDKN2C
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human p18 INK4c. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1031, 12580, 54238
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.