Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDKN3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP158079
Description
CDKN3 Polyclonal specifically detects CDKN3 in Human samples. It is validated for Western Blot.Specifications
CDKN3 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
CDI1KAP1, CDK2-associated dual specificity phosphatase, CDK2-associated dual-specificity phosphatase, Cdk-associated protein phosphatase, CIP2, cyclin-dependent kinase inhibitor 3, cyclin-dependent kinase interacting protein 2, Cyclin-dependent kinase interactor 1, Cyclin-dependent kinase-interacting protein 2, EC 3.1.3.16, EC 3.1.3.48, FLJ25787, KAPMGC70625, Kinase-associated phosphatase | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Equine: 92%; Bovine: 85%; Canine: 85%; Guinea pig: 85%; Mouse: 85%; Pig: 85%; Rat: 85%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q16667 | |
CDKN3 | |
Synthetic peptides corresponding to CDKN3(cyclin-dependent kinase inhibitor 3 (CDK2-associated dual specificity phosphatase)) The peptide sequence was selected from the C terminal of CDKN3. Peptide sequence CKFKDVRRNVQKDTEELKSCGIQDIFVFCTRGELSKYRVPNLLDLYQQCG The peptide sequence for this immunogen was taken from within the described region. | |
100 μL | |
Cell Cycle and Replication, Core ESC Like Genes, Protein Phosphatase, Stem Cell Markers | |
1033 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction