Learn More
CDKN3 Mouse anti-Human, Clone: OTI1H5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503578

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene belongs to the dual specificity protein phosphatase family. It was identified as a cyclin-dependent kinase inhibitor, and has been shown to interact with, and dephosphorylate CDK2 kinase, thus prevent the activation of CDK2 kinase. This gene was reported to be deleted, mutated, or overexpressed in several kinds of cancers. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
CDKN3 | |
Monoclonal | |
Unconjugated | |
CDKN3 | |
CDI1, CIP2, KAP, KAP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1033 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1H5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q16667 | |
CDKN3 | |
Full length human recombit protein of human CDKN3 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.