Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKN3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986765
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6765 Supplier Bioss Supplier No. BS5743R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene belongs to the dual specificity protein phosphatase family. It was identified as a cyclin-dependent kinase inhibitor, and has been shown to interact with, and dephosphorylate CDK2 kinase, thus prevent the activation of CDK2 kinase. This gene was reported to be deleted, mutated, or overexpressed in several kinds of cancers. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen CDKN3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CDKN3
Gene Accession No. Q16667
Gene Alias 2410006H10Rik; CDI1; CDK2-associated dual specificity phosphatase; CDK2-associated dual-specificity phosphatase; CDK2-associated dual-specificity phosphatase {ECO:0000250; Cdk-associated protein phosphatase; CDKN3; cdkn3 {ECO:0000312; CIP2; cyclin dependent kinase inhibitor 3; cyclin-dependent kinase inhibitor; Cyclin-dependent kinase inhibitor 3; cyclin-dependent kinase inhibitor 3 {ECO:0000250; cyclin-dependent kinase interacting protein 2; Cyclin-dependent kinase interactor 1; Cyclin-dependent kinase-interacting protein 2; EMBL:AAI67026.1, ECO:0000312; KAP; KAP1; kinase associated phosphatase; Kinase-associated phosphatase; kinase-associated phosphatase {ECO:0000250; RGD:1307781}; UniProtKB:Q16667}
Gene Symbols CDKN3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CDKN3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1033
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.