Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDMP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987349
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7349 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7349 Supplier Bioss Supplier No. BS6580R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Growth/Differentiation Factor 5 (GDF-5) is a growth factor that regulates cell proliferation and differentiation in embryonic and adult tissues. GDF-5 is part of the TGF family of proteins and is closely related to the BMP family of proteins.

Specifications

Antigen CDMP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GDF5
Gene Accession No. P43026
Gene Alias BDA1C; Bmp14; BMP-14; Bone morphogenetic protein 14; Bp; brachypodism; brp; Cartilage-derived morphogenetic protein 1; cartilage-derived morphogenetic protein-1; CDMP1; CDMP-1; GDF5; Gdf-5; growth differentiation factor 5; Growth/differentiation factor 5; H-GDF-5; LAP4; LAP-4; Lipopolysaccharide-associated protein 4; LPS-associated protein 4; OS5; Radotermin; SYM1B; SYNS2
Gene Symbols GDF5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CDMP1/GDF5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8200
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.