Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDNF Antibody (104), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290322APC
Description
CDNF Monoclonal antibody specifically detects CDNF in Human samples. It is validated for ELISASpecifications
CDNF | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
104 | |
ELISA | |
arginine-rich, mutated in early stage tumors-like 1, ARMETL1, ARMET-like protein 1, cerebral dopamine neurotrophic factor, Conserved dopamine neurotrophic factor | |
Rabbit immunized with purified, recombinant Human CDNF (Accession#: NP_001025125.2, Met1-Leu187) | |
0.1 mL | |
Biologically Active Proteins | |
441549 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction