Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598470
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598470 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598470 Supplier Invitrogen™ Supplier No. PA598470
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDO Polyclonal Antibody for Western Blot, ICC/IF, ELISA

CDON and BOC are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III (FNIII. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDO
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CDON
Gene Accession No. Q4KMG0
Gene Alias CAM-related/down-regulated by oncogenes; Cd0; Cdo; CDON; Cdon homolog; CDON1; cell adhesion associated, oncogene regulated; cell adhesion molecule-related/down-regulated by oncogenes; HPE11; immunoglobulin superfamily member; immunoglobulin superfamily member CDO; oncogene-regulated cell adhesion molecule; ORCAM; surface glycoprotein, Ig superfamily member
Gene Symbols CDON
Host Species Rabbit
Immunogen Recombinant Human Cell adhesion molecule-related/down-regulated by oncogenes protein (1095-1211AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50937
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.