Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDO Recombinant Rabbit Monoclonal Antibody (076), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$293.50 - $565.00
Specifications
Antigen | CDO |
---|---|
Clone | 076 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543462
|
Invitrogen™
MA543462 |
50 μL |
Each for $293.50
|
|
|||||
PIMA543075
|
Invitrogen™
MA543075 |
100 μL |
Each for $565.00
|
|
|||||
Description
Specificity: Human CDON
CDON and BOC are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III (FNIII. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.Specifications
CDO | |
1 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
CAM-related/down-regulated by oncogenes; Cd0; Cdo; CDON; Cdon homolog; CDON1; cell adhesion associated, oncogene regulated; cell adhesion molecule-related/down-regulated by oncogenes; HPE11; immunoglobulin superfamily member; immunoglobulin superfamily member CDO; oncogene-regulated cell adhesion molecule; ORCAM; surface glycoprotein, Ig superfamily member | |
CDON | |
IgG | |
Protein A | |
Antibody |
076 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
Q4KMG0 | |
50937 | |
Recombinant Human CDON protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
CDON |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title