Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA538005
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA538005 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA538005 Supplier Invitrogen™ Supplier No. PA538005
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A recommended positive control is rat liver tissue lysate.

CDO1 (cysteine dioxygenase, type I) belongs to the cysteine dioxygenase family and is involved in organosulfur biosynthesis. CDO1 functions as a dioxygenase that uses iron and zinc as cofactors to catalyze the conversion of L-cysteine and oxygen to 3-sulfinoalanine. CDO1 exists as a monomer and is expressed at high levels in liver and placenta and at lower levels in brain, pancreas and heart. CDO1 is involved in pyruvate-, sulfate- and taurine-related metabolic pathways and is a crucial regulator of cysteine concentrations within the cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDO1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Cdo1
Gene Accession No. P21816, P60334, Q16878
Gene Alias 1300002L19Rik; Cdo; CDO1; CDOI; CDO-I; cysteine dioxygenase 1, cytosolic; cysteine dioxygenase type 1; Cysteine dioxygenase type I; cysteine dioxygenase, type I; cytosolic cysteine dioxygenase 1; D18Ucla3
Gene Symbols Cdo1
Host Species Rabbit
Immunogen 19 amino acid peptide near the amino terminus of human CDO1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1036, 12583, 81718
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.