Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDP-ethanolamine sodium salt hydrate, ≥93% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111003049 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
10 mg
50 mg

Specifications

Linear Formula C11H20N4O11P2 · xNa+ · yH2 O
Synonym Cytidine 5′-diphosphate ethanolamine sodium salt hydrate; Cytidine 5′-diphosphoethanolamine sodium salt hydrate; Cytidine diphosphate ethanolamine sodium salt hydrate
Type Organic Standard
Quantity 50 mg
Formula Weight 446.24 (anhydrous free acid basis)
Percent Purity ≥93% (HPLC)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.