Learn More
Invitrogen™ CDP-Star™ Substrate (12.5 mM Concentrate)
Description
This CDP-Star™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.
- Chemiluminescent substrate provides highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
- Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.
This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours.
Specifications
Specifications
| Content And Storage | Store the product at 4°C. |
| Detection Method | Chemiluminescence |
| Form | Liquid |
| Product Line | CDP-Star, NovaBright |
| Quantity | 5 mL |
| Substrate | Alkaline Phosphatase |
| Substrate Type | AP (Alkaline Phosphatase) Substrate |
| Format | Bottle(s) |
| Product Type | Substrate |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.