Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CDPPB
GREENER_CHOICESDP

Catalog No. 323550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-355-0 50 mg
32-351-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-355-0 Supplier R&D Systems™ Supplier No. 3235/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Positive allosteric modulator of mGlu5 receptors

Brain penetrant, selective positive allosteric modulator at the mGlu5 receptor (EC50 values are 10 and 20 nM for human and rat receptors respectively). Antipsychotic; reverses amphetamine-induced locomotor activity and amphetamine-induced deficits in prepulse inhibition in rats.

Chemical Identifiers

CAS 781652-57-1
Molecular Formula C23H16N4O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 364.4
Purity >98%
Molecular Weight (g/mol) 364.408
Chemical Name or Material CDPPB
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.