Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567271
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567271 Supplier Invitrogen™ Supplier No. PA567271
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEAMDLREDKD FLEDMDSLED MALLEDVDLL EDTDFLEDPD FLEAIDLRED KDFLEDMDSL EDLEAIGRCG F Highest antigen sequence identity to the following orthologs - mouse 57%, rat 33%.

Autoantibodies directed against the protein encoded by CDR1, an intronless gene, have been found in some patients with paraneoplastic cerebellar degeneration. The encoded protein contains several hexapeptide repeats.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDR1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDR1
Gene Accession No. P51861
Gene Alias 100039759; CDR; CDR1; CDR34; CDR62A; cerebellar degeneration related antigen 1; cerebellar degeneration related protein 1; cerebellar degeneration-related antigen 1; cerebellar degeneration-related protein 1, 34kDa; EG631990; Gm2409; Gm7077
Gene Symbols CDR1
Host Species Rabbit
Immunogen Recombinant Human CDR1. Recombinant protein control fragment (Product #RP-107909).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1038
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.