Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568142
Description
Predicted to react with Human samples.
Autoantibodies directed against the protein encoded by CDR1, an intronless gene, have been found in some patients with paraneoplastic cerebellar degeneration. The encoded protein contains several hexapeptide repeats.
Specifications
CDR1 | |
Polyclonal | |
Unconjugated | |
CDR1 | |
100039759; CDR; CDR1; CDR34; CDR62A; cerebellar degeneration related antigen 1; cerebellar degeneration related protein 1; cerebellar degeneration-related antigen 1; cerebellar degeneration-related protein 1, 34kDa; EG631990; Gm2409; Gm7077 | |
Rabbit | |
affinity chromatography | |
RUO | |
1038 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P51861 | |
CDR1 | |
Synthesized peptide derived from human CDR1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction