Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103733
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103733 Supplier Invitrogen™ Supplier No. PA5103733
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CDR1.

Autoantibodies directed against the protein encoded by CDR1, an intronless gene, have been found in some patients with paraneoplastic cerebellar degeneration. The encoded protein contains several hexapeptide repeats.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDR1
Gene Accession No. P51861
Gene Alias 100039759; CDR; CDR1; CDR34; CDR62A; cerebellar degeneration related antigen 1; cerebellar degeneration related protein 1; cerebellar degeneration-related antigen 1; cerebellar degeneration-related protein 1, 34kDa; EG631990; Gm2409; Gm7077
Gene Symbols CDR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDR1(Accession P51861), corresponding to amino acid residues V13-F63.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1038, 631990
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.