Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDR2 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11058345APC
Description
CDR2 Polyclonal antibody specifically detects CDR2 in Human, Mouse, Bovine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
CDR2 | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
CDR62, cerebellar degeneration-related protein (62kD), cerebellar degeneration-related protein 2, cerebellar degeneration-related protein 2, 62kDa, Major Yo paraneoplastic antigen, Paraneoplastic cerebellar degeneration-associated antigen, PCD17, Yo, Yo paraneoplastic antigen | |
A synthetic peptide made to a C-terminal region of human CDR2 [Swiss-Prot# Q01850] | |
0.1 mL | |
Cancer, Hypoxia, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
1039 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Bovine, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction