Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553741 Supplier Invitrogen™ Supplier No. PA553741
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVEKLVPDSL YVPFKEPSQS LLEEMFLTVP ESHRKPLKRS SSETILSSLA GSDIVKGHEE TCIRRAKAVK QRGISLLHEV DTQYSALKVK YEELLKKCQE EQDSLSHKAV QTSRAAAKDL TGV Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Using anti-Yo antibodies, three genes have been cloned (CDR 1-3). CDR2 suppresses the transcriptional activity and DNA binding of nuclear factor-kappa B by an unknown mechanism. The neuron specific expression of CDR2 is regulated by a post-transcriptional mechanism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDR2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDR2
Gene Accession No. Q01850
Gene Alias AA617262; CDR2; CDR62; cerebellar degeneration related protein 2; cerebellar degeneration-related 2; cerebellar degeneration-related protein 2; cerebellar degeneration-related protein 2, 62kDa; major Yo paraneoplastic antigen; MGC128861 protein; paraneoplastic cerebellar degeneration-associated antigen; PCD17; RGD1310578; Yo; Yo paraneoplastic antigen
Gene Symbols CDR2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDR2. Recombinant protein control fragment (Product #RP-91639).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1039
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.