Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDR2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554552
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554552 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554552 Supplier Invitrogen™ Supplier No. PA554552
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSSWRDLRGG EEGQGEVKAG EKSLSQHVEA VDKRLEQSQP EYKALFKEIF SRIQKTKADI NATKVKTHSS Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

The function of CDR2L remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDR2L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cdr2l
Gene Accession No. A2A6T1, Q86X02
Gene Alias CDR2L; cerebellar degeneration related protein 2 like; cerebellar degeneration-related protein 2-like; D030068L24Rik; Gm21; HUMPPA; Paraneoplastic 62 kDa antigen; paraneoplastic antigen; RGD1306881
Gene Symbols Cdr2l
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDR2L. Recombinant protein control fragment (Product #RP-93223).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237988, 30850, 360656
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.