Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDRT15L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571005
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571005 Supplier Thermo Scientific Supplier No. PA571005
Add to Cart
Add to Cart

CDRT15L2 Polyclonal Antibody

CDRT15L2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CDRT15L2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CDRT15L2
Gene Accession No. A8MXV6
Gene Alias CDRT15L2; CMT1A duplicated region transcript 15 protein-like protein; CMT1A duplicated region transcript 15-like 2
Gene Symbols CDRT15L2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human CDRT15L2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256223
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.