Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDRT15L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568393
Description
CDRT15L2 Polyclonal Antibody for IHC (P)
CDRT15L2 is a protein coding gene.
Specifications
CDRT15L2 | |
Polyclonal | |
Unconjugated | |
CDRT15L2 | |
CDRT15L2; CMT1A duplicated region transcript 15 protein-like protein; CMT1A duplicated region transcript 15-like 2 | |
Rabbit | |
affinity chromatography | |
RUO | |
256223 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
A8MXV6 | |
CDRT15L2 | |
Synthetic peptide corresponding to residues near the N terminal of human CDRT15L2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction