Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | CDS1 |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CDS1 Polyclonal antibody specifically detects CDS1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
CDS1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
PBS (pH 7.2) and 40% Glycerol | |
1040 | |
IgG | |
Protein A purified |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
CDP-DAG synthase 1, CDP-DG synthase 1, CDP-DG synthetase 1, CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 1, CDP-diacylglycerol synthase 1, CDP-diglyceride pyrophosphorylase 1, CDP-diglyceride synthase 1, CDP-diglyceride synthetase 1, CDS, CDS 1, CTP:phosphatidate cytidylyltransferase 1, EC 2.7.7.41, phosphatidate cytidylyltransferase 1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: PREAVSPPHREGEAAGGDHETESTSDKETDIDDRYGDLDSRTDSDIPEIPPSSDRTPEILKKALSGLSSR | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title