Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584648 Supplier Invitrogen™ Supplier No. PA584648
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PREAVSPPHR EGEAAGGDHE TESTSDKETD IDDRYGDLDS RTDSDIPEIP PSSDRTPEIL KKALSGLSSR Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 87%.

Breakdown products of phosphoinositides are ubiquitous second messengers that function downstream of many G protein-coupled receptors and tyrosine kinases regulating cell growth, calcium metabolism, and protein kinase C activity. This gene encodes an enzyme which regulates the amount of phosphatidylinositol available for signaling by catalyzing the conversion of phosphatidic acid to CDP-diacylglycerol. This enzyme is an integral membrane protein localized to two subcellular domains, the matrix side of the inner mitochondrial membrane where it is thought to be involved in the synthesis of phosphatidylglycerol and cardiolipin and the cytoplasmic side of the endoplasmic reticulum where it functions in phosphatidylinositol biosynthesis. Two genes encoding this enzyme have been identified in humans, one mapping to human chromosome 4q21 and a second to 20p13. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDS1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDS1
Gene Accession No. Q92903
Gene Alias 4833409J18Rik; AI314024; AW125888; CDP-DAG synthase 1; CDP-DG synthase 1; CDP-DG synthetase 1; CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 1; CDP-diacylglycerol synthase 1; CDP-diglyceride pyrophosphorylase 1; CDP-diglyceride synthase 1; CDP-diglyceride synthetase 1; CDS; CDS 1; Cds1; CTP:phosphatidate cytidylyltransferase 1; phosphatidate cytidylyltransferase 1
Gene Symbols CDS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDS1. Recombinant protein control fragment (Product #RP-108227).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1040
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.