Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDS2 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297196MFV450
Description
CDS2 Polyclonal antibody specifically detects CDS2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CDS2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
CDP-DAG synthase 2, CDP-DG synthase 2, CDP-DG synthetase 2, CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2, CDP-diacylglycerol synthase 2, CDP-diglyceride diphosphorylase 2, CDP-diglyceride pyrophosphorylase 2, CDP-diglyceride synthase 2, CDP-diglyceride synthetase 2, CDS 2, CTP:phosphatidate cytidylyltransferase 2, EC 2.7.7, EC 2.7.7.41, FLJ38111, phosphatidate cytidylyltransferase 2 | |
Produced in rabbits immunized with E. coli-derived Human CDS2 fragment. | |
0.1 mL | |
Endocrinology, Signal Transduction | |
8760 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction