Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDSN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560804 Supplier Invitrogen™ Supplier No. PA560804
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SALPTNDNSY RGILNPSQPG QSSSSSQTFG VSSSGQSVSS NQRPCSSDIP DSPCSGGPIV SHSGPYIPSS HSVSGGQRPV VVVVDQHGSG APG Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 31%.

This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDSN
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDSN
Gene Accession No. Q15517
Gene Alias AI747712; CDSN; Corneodesmosin; D6S586E; differentiated keratinocyte S protein; HTSS; HTSS1; HYPT2; LOC682408; PSS; PSS1; S; S protein; similar to corneodesmosin
Gene Symbols CDSN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDSN. Recombinant protein control fragment (Product #RP-100042).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1041
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.