Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDSN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589932
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589932 Supplier Invitrogen™ Supplier No. PA589932
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGSAGSFKPG TGYSQVSYSS GSGSSLQGAS GSSQLGSSSS HSGSSGSHSG SSSSHSSSSS SFQFSSSSFQ VGNGSALPTN DNSYRGILNP SQPGQSSSSS Q; Positive Samples: BXPC-3, A-431, NIH/3T3, Mouse Skin, Mouse skeletal muscle, Rat skin, Rat skeletal muscle.

This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDSN
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CDSN
Gene Accession No. Q15517, Q7TPC1
Gene Alias AI747712; CDSN; Corneodesmosin; D6S586E; differentiated keratinocyte S protein; HTSS; HTSS1; HYPT2; LOC682408; PSS; PSS1; S; S protein; similar to corneodesmosin
Gene Symbols CDSN
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human CDSN (NP_0012553).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1041, 386463, 682408
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.