Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDT1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501729413 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9413 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9413 Supplier Proteintech Group Inc Supplier No. 143821AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KMT2A is a histone methyltransferase that plays an essential role in early development and hematopoiesis. It is a catalytic subunit of the MLL1/MLL complex, a multiprotein complex that mediates both methylation of ′Lys-4′ of histone H3 (H3K4me) complex and acetylation of ′Lys-16′ of histone H4 (H4K16ac). In the MLL1/MLL complex, KMT2A specifically mediates H3K4me, a tag for epigenetic transcriptional activation. KMT2A has weak methyltransferase activity by itself, and requires other components of the MLL1/MLL complex to obtain full methyltransferase activity. It has no activity toward histone H3 phosphorylated on ′Thr-3′, less activity toward H3 dimethylated on ′Arg-8′ or ′Lys-9′, and has higher activity toward H3 acetylated on ′Lys-9′. Required for transcriptional activation of HOXA9 and promotes PPP1R15A-induced apoptosis.

Specifications

Antigen CDT1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CDT1
Gene Accession No. Q9H211
Gene Alias CDT1, DNA replication factor Cdt1, Double parked homolog, DUP, RIS2
Gene Symbols CDT1
Host Species Rabbit
Immunogen CDT1 Fusion Protein Ag5810
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81620
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.