Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDT2 Antibody (SR2285), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32186150UL
Description
CDT2 Monoclonal antibody specifically detects CDT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
CDT2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
SR2285 | |
Western Blot 1:1000-1:5000, Flow Cytometry 1:80, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:100 | |
CDT2RA-regulated nuclear matrix-associated protein, CDW1, DCAF2DDB1 and CUL4 associated factor 2, DDB1- and CUL4-associated factor 2, denticleless homolog (Drosophila), L2DTLRA regulated nuclear matrix associated protein, Lethal(2) denticleless protein homolog, RAMPdenticleless protein homolog, Retinoic acid-regulated nuclear matrix-associated protein | |
A synthesized peptide derived from human CDT2 (Uniprot #: Q9NZJ0) | |
50 μg | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
51514 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction