Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDV3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556252
Description
Immunogen sequence: LQSTAKHVES RKDKEMEKSF EVVRHKNRGR DEVSKNQALK LQLDNQYAVL ENQKSSHSQY N Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 87%.
The function of this protein remains unknown.
Specifications
CDV3 | |
Polyclonal | |
Unconjugated | |
CDV3 | |
2510010F10Rik; C230084J24Rik; C79446; carnitine deficiency-associated gene expressed in ventricle 3; carnitine deficiency-associated gene expressed in ventricle 3 homolog; carnitine deficiency-associated protein 3; Cdv3; CDV3 homolog; H41; Protein CDV3; protein CDV3 homolog; TPP36; Tyrosine-phosphorylated protein 36 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55573 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKY7 | |
CDV3 | |
Recombinant protein corresponding to Human CDV3. Recombinant protein control fragment (Product #RP-95586). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction