Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDw293 Polyclonal Antibody, Invitrogen™

Catalog No. 409400
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-940-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-940-0 Supplier Invitrogen Supplier No. 409400
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BMPR1B (also known as ALK6, CDw293) is a member of the transmembrane serine/threonine kinase that is the member of the bone morphogenetic protein (BMP) receptor, which is closely related to the activin receptors, ACVR1 and ACVR2. BMPR1B is mainly involved in the endochondral bone formation and embryogenesis. BMPR1B expressed in normal and cancerous prostate tissues and used in the endocrine therapy that given to the prostate cancer patients. BMPR1B receptor trafficking also play a significant role in FOP pathogenesis and used in human T-cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDw293
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene BMPR1B
Gene Accession No. O00238, P36898
Gene Alias Activin receptor-like kinase 6; Acvrlk6; AI385617; Alk6; ALK-6; AMDD; AV355320; BDA1D; BDA2; BMP type-1B receptor; BMP15; Bmpr1b; BMPR-1B; BMPRIB; BMPR-IB; bone morphogenetic protein binding serine/threonine kinase receptor; bone morphogenetic protein receptor IB; bone morphogenetic protein receptor type 1B; bone morphogenetic protein receptor type IB; bone morphogenetic protein receptor type-1B; bone morphogenetic protein receptor, type 1B; bone morphogenetic protein receptor, type IB; CDw293; CFK-43a; serine/threonine receptor kinase; serine/threonine-protein kinase receptor R6; SKR6
Gene Symbols BMPR1B
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of the human, mouse, rat, sheep, pig, and goat BMPR-1B proteins
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12167, 310914, 658
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.